Product Details

SNP ID
rs200796580
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:152709179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAGTGTGCATCCTCATGCCCAC[C/T]TCCAATCTCTTCCTGCTGTGGCTCC
Phenotype
MIM: 612609 MIM: 612618
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCE2A PubMed Links

Gene Details

Gene
LCE2A
Gene Name
late cornified envelope 2A
There are no transcripts associated with this gene.

Gene
LCE4A
Gene Name
late cornified envelope 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178356.3 Intron NP_848133.1

View Full Product Details