Product Details
- SNP ID
-
rs200823619
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:248850497 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCAGGGAAGCCTTCTGGCGGCAGG[G/T]AAACCCGCATATCTCACACCTGGAG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF672
PubMed Links
Gene Details
- Gene
- ZNF672
- Gene Name
- zinc finger protein 672
- Gene
- ZNF692
- Gene Name
- zinc finger protein 692
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136036.2 |
1484 |
Missense Mutation |
ACC,CCC |
T430P |
NP_001129508.1 |
NM_001193328.1 |
1484 |
Missense Mutation |
ACC,CCC |
T380P |
NP_001180257.1 |
NM_017865.3 |
1484 |
Missense Mutation |
ACC,CCC |
T425P |
NP_060335.2 |
XM_011544220.1 |
1484 |
Missense Mutation |
ACC,CCC |
T576P |
XP_011542522.1 |
XM_011544221.1 |
1484 |
Missense Mutation |
ACC,CCC |
T575P |
XP_011542523.1 |
XM_011544222.1 |
1484 |
Missense Mutation |
ACC,CCC |
T425P |
XP_011542524.1 |
XM_011544223.2 |
1484 |
Missense Mutation |
ACC,CCC |
T531P |
XP_011542525.1 |
XM_011544227.1 |
1484 |
Missense Mutation |
ACC,CCC |
T398P |
XP_011542529.1 |
XM_017001698.1 |
1484 |
Missense Mutation |
ACC,CCC |
T424P |
XP_016857187.1 |
XM_017001699.1 |
1484 |
Missense Mutation |
ACC,CCC |
T550P |
XP_016857188.1 |
XM_017001700.1 |
1484 |
Missense Mutation |
ACC,CCC |
T399P |
XP_016857189.1 |
XM_017001701.1 |
1484 |
Missense Mutation |
ACC,CCC |
T424P |
XP_016857190.1 |
XM_017001702.1 |
1484 |
Missense Mutation |
ACC,CCC |
T399P |
XP_016857191.1 |
XM_017001703.1 |
1484 |
Missense Mutation |
ACC,CCC |
T221P |
XP_016857192.1 |
XM_017001704.1 |
1484 |
Missense Mutation |
ACC,CCC |
T221P |
XP_016857193.1 |
XM_017001705.1 |
1484 |
Missense Mutation |
ACC,CCC |
T221P |
XP_016857194.1 |
View Full Product Details