Product Details
- SNP ID
-
rs200200108
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:152109181 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCAGCGGCTGTTCCTCCCTTTCCT[C/G]GAGCAGCTGTTCCTCCTCGCGGAAT
- Phenotype
-
MIM: 190370
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TCHH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72477383] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TCHH
- Gene Name
- trichohyalin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007113.3 |
4131 |
Missense Mutation |
CAG,GAG |
Q1346E |
NP_009044.2 |
View Full Product Details