Product Details
- SNP ID
-
rs200240346
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:182576278 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTTTTTATGCTTTTCTTCATCAA[C/T]GTGTTCTCCCAAATTCCGGATGAAC
- Phenotype
-
MIM: 180435
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNASEL
PubMed Links
Gene Details
- Gene
- RNASEL
- Gene Name
- ribonuclease L
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021133.3 |
2274 |
Missense Mutation |
ATT,GTT |
I673V |
NP_066956.1 |
View Full Product Details