Product Details

SNP ID
rs200933239
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:248180004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATCATGGCATGGGAGAATCAG[A/G]CCTTCAACTCCGACTTCATCCTCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2M2 PubMed Links

Gene Details

Gene
OR2M2
Gene Name
olfactory receptor family 2 subfamily M member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004688.1 19 Missense Mutation ACC,GCC T7A NP_001004688.1

View Full Product Details