Product Details

SNP ID
rs200202023
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:50419212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGGGGGCGCCGCAGGGCCCAGG[C/G]CGGCCGCCAGGCCCCCACGGTGGTG
Phenotype
MIM: 614804
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DMRTA2 PubMed Links

Gene Details

Gene
DMRTA2
Gene Name
DMRT like family A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032110.2 2211 Missense Mutation GCC,GGC A361G NP_115486.1
XM_011541937.2 2211 Missense Mutation GCC,GGC A361G XP_011540239.1

View Full Product Details