Product Details

SNP ID
rs200269153
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:50421243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCGCTCCGCGATGAGCGTGCA[C/T]TTGGCGCACAGGCAGTCCTTCCAGC
Phenotype
MIM: 614804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMRTA2 PubMed Links

Gene Details

Gene
DMRTA2
Gene Name
DMRT like family A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032110.2 1423 Silent Mutation AAA,AAG K98K NP_115486.1
XM_011541937.2 1423 Silent Mutation AAA,AAG K98K XP_011540239.1

View Full Product Details