Product Details
- SNP ID
-
rs200469035
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:209615355 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGCTTCTCCAGTCCTGTCAGGTC[C/T]GCTGAGCGCAGCATGATGGCCTGGC
- Phenotype
-
MIM: 614994
MIM: 150310
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAMK1G
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1049607] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAMK1G
- Gene Name
- calcium/calmodulin dependent protein kinase IG
There are no transcripts associated with this gene.
- Gene
- LAMB3
- Gene Name
- laminin subunit beta 3
- Gene
- MIR4260
- Gene Name
- microRNA 4260
There are no transcripts associated with this gene.
View Full Product Details