Product Details

SNP ID
rs200880274
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:108717890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCTCCTCACGGCTGAAGACGGG[A/G]ACACAGTCATTGAAACCACGGTGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNDC7 PubMed Links

Gene Details

Gene
FNDC7
Gene Name
fibronectin type III domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144937.1 196 Missense Mutation AAC,GAC N66D NP_001138409.1

View Full Product Details