Product Details

SNP ID
rs200283548
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43451305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTAGCCAACTCAAGCCCTCTACT[C/G]TGTCTCCTGCAGGGAGAGGGAGGCC
Phenotype
MIM: 615463
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HYI PubMed Links

Gene Details

Gene
HYI
Gene Name
hydroxypyruvate isomerase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190880.2 963 Missense Mutation ACA,AGA T256R NP_001177809.1
NM_001243526.1 963 UTR 3 NP_001230455.1
NM_031207.5 963 UTR 3 NP_112484.3
XM_005271239.4 963 Missense Mutation ACA,AGA T281R XP_005271296.1
XM_005271240.4 963 Missense Mutation ACA,AGA T247R XP_005271297.1
XM_006710937.3 963 Intron XP_006711000.1
XM_011542222.2 963 UTR 3 XP_011540524.1
Gene
MIR6735
Gene Name
microRNA 6735
There are no transcripts associated with this gene.

Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015284.3 963 UTR 3 NP_056099.3
XM_005270686.3 963 Intron XP_005270743.1
XM_006710501.3 963 Intron XP_006710564.1
XM_011541106.2 963 Intron XP_011539408.1
XM_011541107.2 963 Intron XP_011539409.1
XM_017000819.1 963 Intron XP_016856308.1
XM_017000820.1 963 Intron XP_016856309.1
XM_017000821.1 963 Intron XP_016856310.1

View Full Product Details