Product Details
- SNP ID
-
rs200642266
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:160799895 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGTCCCAAAAATGCTCTTGCTTT[C/T]GCACGTCCCAAAGAAAATGTAACCA
- Phenotype
-
MIM: 600684
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LY9
PubMed Links
Gene Details
- Gene
- LY9
- Gene Name
- lymphocyte antigen 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033667.2 |
381 |
Silent Mutation |
TTC,TTT |
F89F |
NP_001028839.1 |
NM_001261456.1 |
381 |
Silent Mutation |
TTC,TTT |
F89F |
NP_001248385.1 |
NM_001261457.1 |
381 |
Silent Mutation |
TTC,TTT |
F89F |
NP_001248386.1 |
NM_002348.3 |
381 |
Silent Mutation |
TTC,TTT |
F89F |
NP_002339.2 |
XM_011509548.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_011507850.1 |
XM_011509549.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_011507851.1 |
XM_011509550.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_011507852.1 |
XM_011509552.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_011507854.1 |
XM_011509556.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_011507858.1 |
XM_011509560.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_011507862.1 |
XM_017001297.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_016856786.1 |
XM_017001298.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_016856787.1 |
XM_017001299.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_016856788.1 |
XM_017001300.1 |
381 |
Silent Mutation |
TTC,TTT |
F103F |
XP_016856789.1 |
XM_017001301.1 |
381 |
Silent Mutation |
TTC,TTT |
F89F |
XP_016856790.1 |
XM_017001302.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_016856791.1 |
XM_017001303.1 |
381 |
Silent Mutation |
TTC,TTT |
F136F |
XP_016856792.1 |
XM_017001304.1 |
381 |
UTR 5 |
|
|
XP_016856793.1 |
View Full Product Details