Product Details

SNP ID
rs200949626
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:159713557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCTGGGGTTTGGTGAACACTT[A/C]GCCTTGCACTTCATACTTCAGTGCC
Phenotype
MIM: 123260
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRP PubMed Links

Gene Details

Gene
CRP
Gene Name
C-reactive protein, pentraxin-related
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000567.2 747 Missense Mutation NP_000558.2
XM_011509207.2 747 Nonsense Mutation XP_011507509.1

View Full Product Details