Product Details

SNP ID
rs200509955
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:75789340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATGATCCTCATCTTTTATACA[C/G]TCTTAGTGCTGTCCAGGTAAATACT
Phenotype
MIM: 602105 MIM: 179080
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MSH4 PubMed Links

Gene Details

Gene
MSH4
Gene Name
mutS homolog 4
There are no transcripts associated with this gene.

Gene
RABGGTB
Gene Name
Rab geranylgeranyltransferase beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004582.3 371 Missense Mutation ACT,AGT T98S NP_004573.2
Gene
SNORD45A
Gene Name
small nucleolar RNA, C/D box 45A
There are no transcripts associated with this gene.

Gene
SNORD45B
Gene Name
small nucleolar RNA, C/D box 45B
There are no transcripts associated with this gene.

Gene
SNORD45C
Gene Name
small nucleolar RNA, C/D box 45C
There are no transcripts associated with this gene.

View Full Product Details