Product Details
- SNP ID
-
rs200351870
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:212835892 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTACTTTTTGGTGGAAAGTGCTT[C/T]CTCTCCATGTGAGCAAGGAGACTCA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPATA45
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs369495200] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPATA45
- Gene Name
- spermatogenesis associated 45
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001024601.2 |
366 |
Silent Mutation |
AGA,AGG |
R86R |
NP_001019772.1 |
View Full Product Details