Product Details

SNP ID
rs200312696
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:26022863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGAAGATGCCGTTTCACCTCCT[C/G]AAGCCCCTCATGGTGGCAGCTGCAA
Phenotype
MIM: 601738
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXTL1 PubMed Links

Gene Details

Gene
EXTL1
Gene Name
exostosin like glycosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004455.2 1115 Missense Mutation CAA,GAA Q73E NP_004446.2
XM_005245779.3 1115 Missense Mutation CAA,GAA Q73E XP_005245836.1
XM_017000650.1 1115 Missense Mutation CAA,GAA Q73E XP_016856139.1

View Full Product Details