Product Details

SNP ID
rs200576965
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:15440311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGTCTCCCCAGCCTCCAGCTGT[A/G]GGGTGCCCAGCTTCCCGCCCAACCT
Phenotype
MIM: 601405 MIM: 616450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTRC PubMed Links

Gene Details

Gene
CTRC
Gene Name
chymotrypsin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007272.2 75 Missense Mutation AGG,GGG R18G NP_009203.2
XM_011540550.1 75 Missense Mutation AGG,GGG R18G XP_011538852.1
Gene
EFHD2
Gene Name
EF-hand domain family member D2
There are no transcripts associated with this gene.

View Full Product Details