Product Details

SNP ID
rs201877871
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:109250405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCCCTACCACCAGCTCCTGAAG[C/G]CTGCCCCTGGAGCTGTCGCCTCCTG
Phenotype
MIM: 604265
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CELSR2 PubMed Links

Gene Details

Gene
CELSR2
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001408.2 387 Missense Mutation GCC,GGC A109G NP_001399.1
XM_005270580.4 387 Missense Mutation GCC,GGC A109G XP_005270637.1

View Full Product Details