Product Details

SNP ID
rs201878646
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:10403106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATACTGGTGACATCATCATTGA[C/T]GGAGGAAATTCTGAATATAGGGACA
Phenotype
MIM: 172200
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGD PubMed Links

Gene Details

Gene
PGD
Gene Name
phosphogluconate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304451.1 426 Intron NP_001291380.1
NM_001304452.1 426 Silent Mutation GAC,GAT D87D NP_001291381.1
NM_002631.3 426 Silent Mutation GAC,GAT D100D NP_002622.2

View Full Product Details