Product Details

SNP ID
rs201918850
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:1327506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAGGGCCTGCGTACCAGCCCC[A/G]AGGACGCACGCACCTCCGCGCTCTG
Phenotype
MIM: 611354 MIM: 615467 MIM: 601365 MIM: 605865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPSF3L PubMed Links

Gene Details

Gene
CPSF3L
Gene Name
cleavage and polyadenylation specific factor 3-like
There are no transcripts associated with this gene.

Gene
CPTP
Gene Name
ceramide-1-phosphate transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029885.1 803 Missense Mutation AAG,GAG K130E NP_001025056.1
XM_005244801.3 803 Missense Mutation AAG,GAG K130E XP_005244858.1
XM_005244802.1 803 Missense Mutation AAG,GAG K67E XP_005244859.1
XM_011542200.2 803 Missense Mutation AAG,GAG K130E XP_011540502.1
Gene
DVL1
Gene Name
dishevelled segment polarity protein 1
There are no transcripts associated with this gene.

Gene
TAS1R3
Gene Name
taste 1 receptor member 3
There are no transcripts associated with this gene.

View Full Product Details