Product Details

SNP ID
rs201972851
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:203169305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTGGGTGCTGTAGCCAGGGGTG[A/G]AGGTGTGGTCAGCCAGGGGCTGGGT
Phenotype
MIM: 102775 MIM: 601525 MIM: 160795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
There are no transcripts associated with this gene.

Gene
CHI3L1
Gene Name
chitinase 3 like 1
There are no transcripts associated with this gene.

Gene
MYBPH
Gene Name
myosin binding protein H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004997.2 1237 Missense Mutation TCC,TTC S393F NP_004988.2

View Full Product Details