Product Details
- SNP ID
-
rs202196654
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:182575449 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCAGCTCCATCACACTGAGGCTT[A/G]TTTGGACTGTGGGTTTGGGGGAAAT
- Phenotype
-
MIM: 180435
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNASEL
PubMed Links
Gene Details
- Gene
- RNASEL
- Gene Name
- ribonuclease L
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021133.3 |
2426 |
Silent Mutation |
AAC,AAT |
N723N |
NP_066956.1 |
View Full Product Details