Product Details

SNP ID
rs202224828
Assay Type
Functionally Tested
NCBI dbSNP Submissions
11
Location
Chr.1:54807682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCAGGCCATCATGAAAGAGAAG[C/T]GGCTGAAGGTGAGGCCTCTGGGGCC
Phenotype
MIM: 616446
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LEXM PubMed Links
Additional Information
For this assay, SNP(s) [rs671022] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LEXM
Gene Name
lymphocyte expansion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110533.1 403 Missense Mutation CGG,TGG R117W NP_001104003.1
NM_152607.2 403 Missense Mutation CGG,TGG R117W NP_689820.2
XM_017000480.1 403 Missense Mutation CGG,TGG R117W XP_016855969.1
XM_017000481.1 403 Missense Mutation CGG,TGG R117W XP_016855970.1
Gene
TTC22
Gene Name
tetratricopeptide repeat domain 22
There are no transcripts associated with this gene.

View Full Product Details