Product Details

SNP ID
rs202000106
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:39738921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGATGGCCACCACCAAGCGCGT[C/T]TTGTACGTGGGTGAGCAGGAGGGGT
Phenotype
MIM: 602435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPIE PubMed Links

Gene Details

Gene
PPIE
Gene Name
peptidylprolyl isomerase E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195007.1 77 Intron NP_001181936.1
NM_001319293.1 77 Intron NP_001306222.1
NM_006112.3 77 Silent Mutation GTC,GTT V7V NP_006103.1
NM_203456.2 77 Intron NP_982281.1
XM_006710289.2 77 Intron XP_006710352.1
XM_006710290.3 77 Silent Mutation GTC,GTT V7V XP_006710353.1
XM_011540501.1 77 Intron XP_011538803.1
XM_017000051.1 77 Intron XP_016855540.1
XM_017000052.1 77 UTR 5 XP_016855541.1

View Full Product Details