Product Details

SNP ID
rs202039790
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:203128978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCGCTGCGGGATGCCACCTTCT[A/G]CTTCATCGTGTCGCTGGCGGTGGCT
Phenotype
MIM: 102775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000674.2 65 Missense Mutation TAC,TGC Y46C NP_000665.1
NM_001048230.1 65 Missense Mutation TAC,TGC Y46C NP_001041695.1
XM_005244901.1 65 UTR 5 XP_005244958.1
XM_005244902.4 65 UTR 5 XP_005244959.1
XM_017000342.1 65 UTR 5 XP_016855831.1

View Full Product Details