Product Details

SNP ID
rs201963397
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:15114516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGGCTTCAGCAGCAAAGATCCC[A/G]ATTTCCATGATGACTATGGCTCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAZN PubMed Links

Gene Details

Gene
KAZN
Gene Name
kazrin, periplakin interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017999.2 4482 Intron NP_001017999.1
NM_001018000.3 4482 Intron NP_001018000.1
NM_001018001.2 4482 Intron NP_001018001.1
NM_015209.2 4482 Intron NP_056024.1
NM_201628.2 4482 Missense Mutation AAT,GAT N737D NP_963922.2
XM_005245795.4 4482 Missense Mutation AAT,GAT N835D XP_005245852.1
XM_011541074.2 4482 UTR 3 XP_011539376.1
XM_011541075.2 4482 UTR 3 XP_011539377.1
XM_011541076.2 4482 UTR 3 XP_011539378.1
XM_011541077.2 4482 UTR 3 XP_011539379.1
XM_011541080.2 4482 Intron XP_011539382.1
XM_011541081.2 4482 UTR 3 XP_011539383.1
XM_017000768.1 4482 Intron XP_016856257.1
XM_017000769.1 4482 Intron XP_016856258.1
XM_017000770.1 4482 Intron XP_016856259.1
XM_017000771.1 4482 Intron XP_016856260.1
XM_017000772.1 4482 Intron XP_016856261.1
Gene
TMEM51-AS1
Gene Name
TMEM51 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details