Product Details

SNP ID
rs202080866
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:3781165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTACACCACCTTCAGGCTCCCTT[C/T]CAGATCCACCACCCGGACCTGCTCC
Phenotype
MIM: 615242
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC27 PubMed Links

Gene Details

Gene
CCDC27
Gene Name
coiled-coil domain containing 27
There are no transcripts associated with this gene.

Gene
LRRC47
Gene Name
leucine rich repeat containing 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020710.2 1703 Missense Mutation AAA,GAA K559E NP_065761.1
Gene
SMIM1
Gene Name
small integral membrane protein 1 (Vel blood group)
There are no transcripts associated with this gene.

View Full Product Details