Product Details

SNP ID
rs202068651
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:15573706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCATCTCAAACAGCAGGTTAGCC[A/G]CCAGCAGGGCTGTGTTCCCTAAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGMAT PubMed Links

Gene Details

Gene
AGMAT
Gene Name
agmatinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024758.4 1147 Missense Mutation GCG,GTG A335V NP_079034.3
Gene
DNAJC16
Gene Name
DnaJ heat shock protein family (Hsp40) member C16
There are no transcripts associated with this gene.

View Full Product Details