Product Details

SNP ID
rs201832076
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:248180029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTCAACTCCGACTTCATCCTCC[C/T]TGGAATCTTCAATCACAGCCCACCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2M2 PubMed Links

Gene Details

Gene
OR2M2
Gene Name
olfactory receptor family 2 subfamily M member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004688.1 44 Missense Mutation CCT,CTT P15L NP_001004688.1

View Full Product Details