Product Details

SNP ID
rs202161529
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:31182575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACTTTGCTCACGTAGCAGGCGA[A/T]CACGAAGCCGAACAGCTGGGAGTGG
Phenotype
MIM: 612871
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NKAIN1 PubMed Links

Gene Details

Gene
NKAIN1
Gene Name
Na+/K+ transporting ATPase interacting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024522.2 827 Missense Mutation ATC,TTC I163F NP_078798.2
XM_017002320.1 827 Missense Mutation ATC,TTC I136F XP_016857809.1

View Full Product Details