Product Details

SNP ID
rs202083240
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:35888449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTGCGGGCGCTTACCTGCCCCC[A/C]CTGCAGCAGGTGTTCCAGGCACCTC
Phenotype
MIM: 606228
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AGO1 PubMed Links

Gene Details

Gene
AGO1
Gene Name
argonaute 1, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317122.1 236 Silent Mutation CCA,CCC P16P NP_001304051.1
NM_001317123.1 236 UTR 5 NP_001304052.1
NM_012199.4 236 Silent Mutation CCA,CCC P16P NP_036331.1
XM_011541236.2 236 Silent Mutation CCA,CCC P16P XP_011539538.1
XM_017001016.1 236 Silent Mutation CCA,CCC P16P XP_016856505.1
XM_017001017.1 236 Silent Mutation CCA,CCC P16P XP_016856506.1
XM_017001018.1 236 Intron XP_016856507.1
XM_017001019.1 236 UTR 5 XP_016856508.1

View Full Product Details