Product Details
- SNP ID
-
rs201886827
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:241497928 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAGTTTCCTTTAAGGTTGATCCA[C/T]TTTTGTGTGCTGTCTTAGCAATCTT
- Phenotype
-
MIM: 136850
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FH
PubMed Links
Gene Details
- Gene
- FH
- Gene Name
- fumarate hydratase
View Full Product Details