Product Details

SNP ID
rs202045380
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:156595485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGAGGGACTGATTGGTTTGCT[C/G]TCTGCTATTCAATTCCCCAAGCCCA
Phenotype
MIM: 608862
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPATCH4 PubMed Links

Gene Details

Gene
GPATCH4
Gene Name
G-patch domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015590.3 900 Missense Mutation CAG,GAG Q286E NP_056405.2
NM_182679.2 900 Missense Mutation CAG,GAG Q281E NP_872620.1
XM_005245287.3 900 Missense Mutation CAG,GAG Q252E XP_005245344.1
Gene
HAPLN2
Gene Name
hyaluronan and proteoglycan link protein 2
There are no transcripts associated with this gene.

Gene
NAXE
Gene Name
NAD(P)HX epimerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144772.2 900 Intron NP_658985.2
XM_017000319.1 900 Intron XP_016855808.1
Gene
TTC24
Gene Name
tetratricopeptide repeat domain 24
There are no transcripts associated with this gene.

View Full Product Details