Product Details

SNP ID
rs202084179
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:1211615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTCTTGGATGGGGGTCCGGAA[A/G]CTGCCTCCCCCTGGGGAGGAAAAAA
Phenotype
MIM: 614282 MIM: 603905 MIM: 600315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDF4 PubMed Links

Gene Details

Gene
SDF4
Gene Name
stromal cell derived factor 4
There are no transcripts associated with this gene.

Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
There are no transcripts associated with this gene.

Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003327.3 1247 Silent Mutation AGC,AGT S258S NP_003318.1
XM_011542074.2 1247 Silent Mutation AGC,AGT S288S XP_011540376.1
XM_011542075.2 1247 Silent Mutation AGC,AGT S284S XP_011540377.1
XM_011542076.2 1247 Silent Mutation AGC,AGT S262S XP_011540378.1
XM_011542077.2 1247 Silent Mutation AGC,AGT S184S XP_011540379.1
XM_017002231.1 1247 Silent Mutation AGC,AGT S284S XP_016857720.1
XM_017002232.1 1247 Silent Mutation AGC,AGT S258S XP_016857721.1

View Full Product Details