Product Details

SNP ID
rs202151018
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:23193565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTTCCGGGCAGCCCGGTAGATCC[A/G]GCCATATAGGATGATGAGCAACACC
Phenotype
MIM: 182133
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR1D PubMed Links

Gene Details

Gene
HTR1D
Gene Name
5-hydroxytryptamine receptor 1D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000864.4 1165 Missense Mutation CGG,TGG R219W NP_000855.1

View Full Product Details