Product Details
- SNP ID
-
rs200056526
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:56004091 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACGATGGGCTCCGTGTCGTTCTG[C/T]GCCCAGACGGGCAGCCCCGGCAGCG
- Phenotype
-
MIM: 615029
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBLN4
PubMed Links
Gene Details
- Gene
- CBLN4
- Gene Name
- cerebellin 4 precursor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080617.5 |
1382 |
Silent Mutation |
GCA,GCG |
A27A |
NP_542184.1 |
View Full Product Details