Product Details

SNP ID
rs201222711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:21512139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGACCAAGACGGGCACGGCCAC[A/C]CGGCGCGGCGAGGGCAGGGGCGTCA
Phenotype
MIM: 604612
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NKX2-2 PubMed Links

Gene Details

Gene
NKX2-2
Gene Name
NK2 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002509.3 620 Silent Mutation CGG,CGT R202R NP_002500.1
XM_006723566.3 620 Silent Mutation CGG,CGT R131R XP_006723629.1

View Full Product Details