Product Details

SNP ID
rs202166182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58840352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTATTCCCTGAGTCCCCCGAATC[A/G]GAATCTGACCACGAGCACGAGGAGG
Phenotype
MIM: 139320 MIM: 610540
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAS PubMed Links

Gene Details

Gene
GNAS
Gene Name
GNAS complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000516.5 672 Intron NP_000507.1
NM_001077488.3 672 Intron NP_001070956.1
NM_001077489.3 672 Intron NP_001070957.1
NM_001077490.2 672 Intron NP_001070958.1
NM_001309840.1 672 Intron NP_001296769.1
NM_001309842.1 672 Intron NP_001296771.1
NM_001309861.1 672 Intron NP_001296790.1
NM_001309883.1 672 Intron NP_001296812.1
NM_016592.3 672 Silent Mutation TCA,TCG S82S NP_057676.1
NM_080425.3 672 Intron NP_536350.2
NM_080426.3 672 Intron NP_536351.1
XM_017027812.1 672 Intron XP_016883301.1
XM_017027813.1 672 Intron XP_016883302.1
XM_017027814.1 672 Intron XP_016883303.1
XM_017027815.1 672 Intron XP_016883304.1
XM_017027816.1 672 Intron XP_016883305.1
XM_017027817.1 672 Intron XP_016883306.1
XM_017027818.1 672 Intron XP_016883307.1
XM_017027819.1 672 Intron XP_016883308.1
XM_017027820.1 672 Intron XP_016883309.1
XM_017027821.1 672 Silent Mutation TCA,TCG S82S XP_016883310.1
XM_017027822.1 672 Silent Mutation TCA,TCG S82S XP_016883311.1
Gene
GNAS-AS1
Gene Name
GNAS antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details