Product Details

SNP ID
rs200264938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38969159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCTGGGACAACGGTTGTTTACA[A/G]CCCTTATGCTGCCCTTTCCATAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX35 PubMed Links

Gene Details

Gene
DHX35
Gene Name
DEAH-box helicase 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190809.1 149 Missense Mutation AAC,AGC N40S NP_001177738.1
NM_021931.3 149 Missense Mutation AAC,AGC N40S NP_068750.2
XM_006723848.3 149 Intron XP_006723911.1
XM_011528972.1 149 Missense Mutation AAC,AGC N40S XP_011527274.1
XM_011528974.1 149 Missense Mutation AAC,AGC N40S XP_011527276.1
XM_017028004.1 149 Intron XP_016883493.1

View Full Product Details