Product Details
- SNP ID
-
rs200463772
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:59866509 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATTACAGACCTAGAGTCCTGACT[G/T]TGATGATTCATTGTTCTCAGATGTT
- Phenotype
-
MIM: 604105
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SYCP2
PubMed Links
Gene Details
- Gene
- SYCP2
- Gene Name
- synaptonemal complex protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014258.3 |
4391 |
Missense Mutation |
CAA,CAC |
Q1402H |
NP_055073.2 |
XM_011528487.2 |
4391 |
Missense Mutation |
CAA,CAC |
Q1409H |
XP_011526789.1 |
XM_011528488.2 |
4391 |
Missense Mutation |
CAA,CAC |
Q1409H |
XP_011526790.1 |
XM_011528489.2 |
4391 |
Missense Mutation |
CAA,CAC |
Q1402H |
XP_011526791.1 |
XM_011528490.2 |
4391 |
Missense Mutation |
CAA,CAC |
Q1401H |
XP_011526792.1 |
XM_011528493.2 |
4391 |
Missense Mutation |
CAA,CAC |
Q1237H |
XP_011526795.1 |
XM_017027586.1 |
4391 |
Missense Mutation |
CAA,CAC |
Q1409H |
XP_016883075.1 |
XM_017027587.1 |
4391 |
Missense Mutation |
CAA,CAC |
Q1408H |
XP_016883076.1 |
XM_017027588.1 |
4391 |
Missense Mutation |
CAA,CAC |
Q1357H |
XP_016883077.1 |
XM_017027589.1 |
4391 |
Missense Mutation |
CAA,CAC |
Q1352H |
XP_016883078.1 |
XM_017027590.1 |
4391 |
Missense Mutation |
CAA,CAC |
Q1237H |
XP_016883079.1 |
XM_017027591.1 |
4391 |
Missense Mutation |
CAA,CAC |
Q926H |
XP_016883080.1 |
XM_017027592.1 |
4391 |
Intron |
|
|
XP_016883081.1 |
View Full Product Details