Product Details

SNP ID
rs200476717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18489964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGGCAGTTGGCCTTGATCTTCT[C/T]CCACTGCCAGGGGCGGGTGAAGTAT
Phenotype
MIM: 602908
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR3F PubMed Links

Gene Details

Gene
POLR3F
Gene Name
RNA polymerase III subunit F
There are no transcripts associated with this gene.

Gene
RBBP9
Gene Name
RB binding protein 9, serine hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006606.2 437 Missense Mutation AAG,GAG K121E NP_006597.2
XM_005260652.2 437 Missense Mutation AAG,GAG K63E XP_005260709.1

View Full Product Details