Product Details

SNP ID
rs200631954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45834265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTGGCCGGCGACGGCCCCGGGG[A/G]GGCGGCGGCCAGTCCAGAGGCCGAG
Phenotype
MIM: 608123 MIM: 191039
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACOT8 PubMed Links

Gene Details

Gene
ACOT8
Gene Name
acyl-CoA thioesterase 8
There are no transcripts associated with this gene.

Gene
SNX21
Gene Name
sorting nexin family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042632.2 175 Missense Mutation GAG,GGG E29G NP_001036097.1
NM_001042633.2 175 Missense Mutation GAG,GGG E29G NP_001036098.1
NM_033421.3 175 Missense Mutation GAG,GGG E29G NP_219489.1
NM_152897.2 175 Missense Mutation GAG,GGG E29G NP_690857.1
XM_005260608.4 175 Intron XP_005260665.1
XM_011529095.2 175 Intron XP_011527397.1
XM_011529096.2 175 Missense Mutation GAG,GGG E29G XP_011527398.1
XM_011529098.2 175 Missense Mutation GAG,GGG E29G XP_011527400.1
XM_011529099.2 175 Missense Mutation GAG,GGG E29G XP_011527401.1
XM_017028120.1 175 Missense Mutation GAG,GGG E29G XP_016883609.1
Gene
TNNC2
Gene Name
troponin C2, fast skeletal type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003279.2 175 Intron NP_003270.1
XM_011529031.2 175 Intron XP_011527333.1

View Full Product Details