Product Details

SNP ID
rs202088044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36612157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCCCCTTGGAACTTGGAAGA[C/T]CCGTGTTTCCTGGACCGCGAATCAG
Phenotype
MIM: 606577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf24 PubMed Links

Gene Details

Gene
C20orf24
Gene Name
chromosome 20 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199534.1 690 UTR 3 NP_001186463.1
NM_018840.4 690 UTR 3 NP_061328.1
NM_199483.2 690 Silent Mutation GAC,GAT D122D NP_955777.1
Gene
SLA2
Gene Name
Src like adaptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032214.3 690 Intron NP_115590.1
NM_175077.2 690 Intron NP_778252.1
XM_017028098.1 690 Intron XP_016883587.1
Gene
TGIF2-C20orf24
Gene Name
TGIF2-C20orf24 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199535.1 690 UTR 3 NP_001186464.1

View Full Product Details