Product Details

SNP ID
rs201045858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45375116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGGCGCTTGACCCAACGCAGGG[G/T]GGAGGATCTGCACATCACCCTGGGC
Phenotype
MIM: 612979
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SYS1 PubMed Links

Gene Details

Gene
SYS1
Gene Name
Sys1 golgi trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099791.2 1031 UTR 3 NP_001093261.1
NM_001197129.1 1031 Intron NP_001184058.1
NM_033542.3 1031 Intron NP_291020.1
Gene
SYS1-DBNDD2
Gene Name
SYS1-DBNDD2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TP53TG5
Gene Name
TP53 target 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014477.2 1031 Missense Mutation ACC,CCC T231P NP_055292.1
XM_011528790.2 1031 Missense Mutation ACC,CCC T215P XP_011527092.1

View Full Product Details