Product Details
- SNP ID
-
rs200792247
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23444044 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGCAAAGCAAGAAGCTGAGAAA[C/G]GTGTGTAGTGGAAGTCATCCCAAAG
- Phenotype
-
MIM: 609731
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CST11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12625959] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CST11
- Gene Name
- cystatin 11
There are no transcripts associated with this gene.
- Gene
- CSTL1
- Gene Name
- cystatin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138283.1 |
576 |
Missense Mutation |
AAC,AAG |
N110K |
NP_612140.1 |
View Full Product Details