Product Details

SNP ID
rs200089008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43673840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGAGCTGCACTACCAGGACACA[C/G]ATTCAGATGTGCCGGAGCAGAGGGA
Phenotype
MIM: 601415
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYBL2 PubMed Links

Gene Details

Gene
MYBL2
Gene Name
MYB proto-oncogene like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278610.1 320 Missense Mutation CAT,GAT H19D NP_001265539.1
NM_002466.3 320 Missense Mutation CAT,GAT H19D NP_002457.1

View Full Product Details