Product Details

SNP ID
rs200193714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:16732221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTTGGTCATGTGGTGGACATTG[A/T]GGCTTTAAAGACCATGAAGATGAGG
Phenotype
MIM: 603520
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SNRPB2 PubMed Links

Gene Details

Gene
SNRPB2
Gene Name
small nuclear ribonucleoprotein polypeptide B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003092.4 258 Missense Mutation GAG,GTG E41V NP_003083.1
NM_198220.2 258 Missense Mutation GAG,GTG E41V NP_937863.1

View Full Product Details