Product Details

SNP ID
rs201051836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56249115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGACATGCTGGTAAGTGTGTCCA[A/G]TGCCCTGGAGACCATCATGATCGCC
Phenotype
MIM: 155540
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MC3R PubMed Links

Gene Details

Gene
MC3R
Gene Name
melanocortin 3 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019888.3 384 Missense Mutation AAT,AGT N91S NP_063941.3

View Full Product Details