Product Details
- SNP ID
-
rs200506017
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23826508 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTGGCCTTGTTATACTCGCTGAT[G/T]ACAAAGTGAAGGGCACGCTGTACCC
- Phenotype
-
MIM: 123856
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CST2
PubMed Links
Gene Details
- Gene
- CST2
- Gene Name
- cystatin SA
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001322.2 |
168 |
Silent Mutation |
GTA,GTC |
V51V |
NP_001313.1 |
View Full Product Details