Product Details

SNP ID
rs201519741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34559894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGCCTCCCAGGAAACCTTCGGCT[G/T]CTGAGCCAGCAGTAGGGGGGCTCGG
Phenotype
MIM: 601242 MIM: 608528
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAP1LC3A PubMed Links

Gene Details

Gene
MAP1LC3A
Gene Name
microtubule associated protein 1 light chain 3 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032514.3 835 Missense Mutation TGC,TTC C121F NP_115903.1
NM_181509.2 835 Missense Mutation TGC,TTC C125F NP_852610.1
XM_011529083.2 835 Missense Mutation TGC,TTC C223F XP_011527385.1
XM_011529084.2 835 Missense Mutation TGC,TTC C223F XP_011527386.1
XM_011529085.2 835 Missense Mutation TGC,TTC C182F XP_011527387.1
Gene
PIGU
Gene Name
phosphatidylinositol glycan anchor biosynthesis class U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080476.4 835 Intron NP_536724.1
XM_011528542.2 835 Intron XP_011526844.1
XM_017027664.1 835 Intron XP_016883153.1
XM_017027665.1 835 Intron XP_016883154.1

View Full Product Details