Product Details

SNP ID
rs199502068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36607785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTCCTGGGTGTCATTTGGGGAG[G/T]TTTGCCATTACGAGGGTTCTTGGGA
Phenotype
MIM: 606577
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C20orf24 PubMed Links

Gene Details

Gene
C20orf24
Gene Name
chromosome 20 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199534.1 379 Missense Mutation GGT,GTT G62V NP_001186463.1
NM_018840.4 379 Missense Mutation GGT,GTT G62V NP_061328.1
NM_199483.2 379 Missense Mutation GGT,GTT G62V NP_955777.1
Gene
SLA2
Gene Name
Src like adaptor 2
There are no transcripts associated with this gene.

Gene
TGIF2-C20orf24
Gene Name
TGIF2-C20orf24 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199535.1 379 Missense Mutation GGT,GTT G88V NP_001186464.1

View Full Product Details